Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001291892.1 | NP_001278821.1 | 3 | 361 | |
NM_008147.2 | NP_032173.1 | 3 | 356 | ||
GenBank mRNA | AK155788.1 | - | 1 | 611 | |
AK167751.1 | - | 3 | 277 | ||
AK170384.1 | - | 3 | 293 | ||
AK170836.1 | - | 2 | 499 | ||
AK171133.1 | - | 3 | 298 | ||
AK171229.1 | - | 3 | 298 | ||
AK171573.1 | - | 3 | 305 | ||
BY204344.1 | - | 3 | 232 | ||
M65027.1 | - | 3 | 289 |