Gene Transcripts

Gene Symbol
Sprr2f
Entrez Gene ID
20760
Gene Name
small proline-rich protein 2F
Gene Aliases
Chromosome Location
Chr. 3: 92365187 - 92366442 on Build GRCm38
UniGene ID
Mm.10692
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011472.2 NP_035602.1 2 303
GenBank mRNA AJ005564.1 - 1 264
AY158990.1 - 2 303
BC120756.1 - 1 272
BC120758.1 - 1 272

View Full Product Details