Gene Transcripts
- Gene Symbol
- Xlr
- Entrez Gene ID
- 22441
- Gene Name
- X-linked lymphocyte-regulated
- Gene Aliases
- Chromosome Location
- Chr. X: 53777118 - 53797716 on Build GRCm38
- UniGene ID
- Mm.379041
- Species
- Mus musculus
- Gene Symbol
- 3830403N18Rik
- Entrez Gene ID
- 70691
- Gene Name
- RIKEN cDNA 3830403N18 gene
- Gene Aliases
- Xlrl
- Chromosome Location
- Chr. X: 55839252 - 56153496 on Build GRCm38
- UniGene ID
- Mm.335676
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK014417.1 |
- |
4 |
357 |
|
View Full Product Details