Gene Transcripts

Gene Symbol
Mxd3
Entrez Gene ID
17121
Gene Name
Max dimerization protein 3
Gene Aliases
4631412E13Rik, Mad3, bHLHc13
Chromosome Location
Chr. 13: 55325168 - 55329770 on Build GRCm38
UniGene ID
Mm.20350
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016662.4 NP_057871.2 4 293
GenBank mRNA AK086190.1 - 2 1049
AK138742.1 - 4 294
BC051970.1 - 4 296 5714980
U32394.1 - 4 230

View Full Product Details