Gene Transcripts

Gene Symbol
Mrpl53
Entrez Gene ID
68499
Gene Name
mitochondrial ribosomal protein L53
Gene Aliases
1110007K17Rik
Chromosome Location
Chr. 6: 83109108 - 83109932 on Build GRCm38
UniGene ID
Mm.333183
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026744.3 NP_081020.1 2, 3 220
GenBank mRNA AK003549.1 - 2, 3 221
BC022162.1 - 2, 3 245 5148541
BC065995.1 - 1443 5688315

View Full Product Details