Gene Transcripts

Gene Symbol
Pi15
Entrez Gene ID
94227
Gene Name
peptidase inhibitor 15
Gene Aliases
P24TI, P25TI, PI-15, SugarCrisp
Chromosome Location
Chr. 1: 17601901 - 17630939 on Build GRCm38
UniGene ID
Mm.442452
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053191.2 NP_444421.2 2 285
GenBank mRNA AF329196.1 - 2 205
AK030960.1 - 2 286
AK040961.1 - 2 284
BC116705.1 - 2 181 40102877

View Full Product Details