Gene Transcripts

Gene Symbol
Mrpl33
Entrez Gene ID
66845
Gene Name
mitochondrial ribosomal protein L33
Gene Aliases
0610009M10Rik, L33mt, MRP-L33
Chromosome Location
Chr. 5: 31613951 - 31622644 on Build GRCm38
UniGene ID
Mm.120763
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025796.3 NP_080072.2 4 235
GenBank mRNA AB049651.1 - 4 218
AK002421.1 - 4 215
AK008431.1 - 4 215
AK021173.1 - 4 215
AK041771.1 - 2 2513
AK167883.1 - 4 215
BC006029.1 - 2 2499
BC037363.1 - 4 201 1514355
BC051471.1 - 4 212 1514101
BC055724.1 - 4 194 6411227
BC059722.1 - 4 214 6772871
BY112771.1 - 4 235

View Full Product Details