Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028967.1 | NP_083243.1 | 2, 3 | 212 | |
GenBank mRNA | AK016990.1 | - | 428 | ||
AK150133.1 | - | 2, 3 | 232 | ||
AK171828.1 | - | 2, 3 | 229 | ||
BC024521.1 | - | 2, 3 | 212 | 4984403 |