Gene Transcripts

Gene Symbol
Klra22
Entrez Gene ID
93969
Gene Name
killer cell lectin-like receptor subfamily A, member 22
Gene Aliases
Ly49v
Chromosome Location
Chr. : - on Build
UniGene ID
Mm.333431
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053152.2 NP_444382.1 7 1064
GenBank mRNA AF288381.1 - 7 1064

View Full Product Details