Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145216.3 | NP_660251.1 | 2 | 485 | |
GenBank mRNA | AF498251.1 | - | 2 | 307 | |
AK008807.1 | - | 2 | 486 | ||
AK044011.1 | - | 2 | 486 | ||
BC107200.1 | - | 3 | 368 | ||
BC107201.1 | - | 3 | 366 | 40051192 |