Gene Transcripts

Gene Symbol
Slc35c1
Entrez Gene ID
228368
Gene Name
solute carrier family 35, member C1
Gene Aliases
E430007K15Rik, fuct1
Chromosome Location
Chr. 2: 92452764 - 92460518 on Build GRCm38
UniGene ID
Mm.273990
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145832.3 NP_665831.1 2 605
NM_211358.2 NP_997597.1 1 1093
GenBank mRNA AJ440720.1 - 1 490
AK030977.1 - 2 606
AK033295.1 - 1 182
AK040986.1 - 1 840
AK045972.1 - 2 582
AK050311.1 - 2 570
AK081068.1 - 1 591
AK088227.1 - 2 592
AK138595.1 - 1 1036
AK144762.1 - 1 1015
AK151154.1 - 1 1089
BC031814.1 - 1 687 4196818
BC116736.1 - 1 518 40126055
BC119128.1 - 1 518 8733877

View Full Product Details