Gene Transcripts

Gene Symbol
PBK
Entrez Gene ID
55872
Gene Name
PDZ binding kinase
Gene Aliases
CT84, HEL164, Nori-3, SPK, TOPK
Chromosome Location
Chr. 8: 27809621 - 27838095 on Build GRCh38
UniGene ID
Hs.104741
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278945.1 NP_001265874.1 4 577
NM_018492.3 NP_060962.2 4 635
XM_006716368.2 XP_006716431.1 4 559
GenBank mRNA AB027249.1 - 4 361
AB027250.1 - 4 341
AF189722.1 - 4 325
AF237709.1 - 4 349
AK027291.1 - 4 340
AK301836.1 - 4 537
BC015191.1 - 4 349 4082846
BI461026.1 - 4 378
EU794685.1 - 4 372

View Full Product Details