Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256139.1 | NP_001243068.1 | 5 | 467 | |
NM_001256140.1 | NP_001243069.1 | 5 | 757 | ||
NM_001320732.1 | NP_001307661.1 | 5 | 443 | ||
NM_001320733.1 | NP_001307662.1 | 5 | 404 | ||
NM_001320734.1 | NP_001307663.1 | 5 | 757 | ||
NM_001747.3 | NP_001738.2 | 5 | 757 | ||
XM_011533122.1 | XP_011531424.1 | 5 | 577 | ||
XM_011533123.1 | XP_011531425.1 | 5 | 473 | ||
GenBank mRNA | AK130130.1 | - | 5 | 757 | |
AK225374.1 | - | 5 | 430 | ||
AK225378.1 | - | 5 | 430 | ||
AK300507.1 | - | 5 | 349 | ||
AK309290.1 | - | 4 | 126 | ||
AK313898.1 | - | 5 | 411 | ||
BC000728.2 | - | 5 | 411 | 3505088 | |
BC014549.1 | - | 5 | 413 | 4108934 | |
BG762114.1 | - | 5 | 405 | ||
BG762391.1 | - | 5 | 443 | ||
BU172582.1 | - | 5 | 409 | ||
FJ224312.1 | - | 5 | 613 | ||
M94345.1 | - | 5 | 412 |