Gene Transcripts

Gene Symbol
Hdhd3
Entrez Gene ID
72748
Gene Name
haloacid dehalogenase-like hydrolase domain containing 3
Gene Aliases
2810435D12Rik
Chromosome Location
Chr. 4: 62499054 - 62502200 on Build GRCm38
UniGene ID
Mm.41506
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024257.1 NP_077219.1 2 319
GenBank mRNA AK013245.1 - 2 334
BC003491.1 - 2 319 2811559

View Full Product Details