Gene Transcripts

Gene Symbol
EPHB4
Entrez Gene ID
2050
Gene Name
EPH receptor B4
Gene Aliases
HTK, MYK1, TYRO11
Chromosome Location
Chr. 7: 100802565 - 100827521 on Build GRCh38
UniGene ID
Hs.437008
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004444.4 NP_004435.3 14 2837
XM_017011816.1 XP_016867305.1 14 2494
GenBank mRNA AB209644.1 - 13 3547
AK301224.1 - 9 1144
AY056048.1 - 14 2431
BC004264.1 - 11 1922
BC052804.1 - 14 2514 6584384
U07695.1 - 14 2431

View Full Product Details