Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001203.2 | NP_001194.1 | 7 | 659 | |
NM_001256792.1 | NP_001243721.1 | 5 | 555 | ||
NM_001256793.1 | NP_001243722.1 | 5 | 496 | ||
NM_001256794.1 | NP_001243723.1 | 5 | 539 | ||
XM_011532201.2 | XP_011530503.1 | 6 | 538 | ||
XM_017008558.1 | XP_016864047.1 | 7 | 730 | ||
XM_017008559.1 | XP_016864048.1 | 8 | 720 | ||
XM_017008560.1 | XP_016864049.1 | 5 | 612 | ||
XM_017008561.1 | XP_016864050.1 | 6 | 569 | ||
GenBank mRNA | AK293008.1 | - | 5 | 539 | |
AK299930.1 | - | 5 | 496 | ||
AK313642.1 | - | 5 | 555 | ||
BC047773.1 | - | 4 | 455 | 6202951 | |
BC069796.1 | - | 6 | 571 | 7262484 | |
BC069803.1 | - | 6 | 571 | 7262496 | |
D89675.1 | - | 4 | 403 | ||
U89326.1 | - | 7 | 658 |