Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_174922.4 | NP_777582.4 | 5, 6 | 629 | |
XM_006716527.2 | XP_006716590.1 | 5, 6 | 619 | ||
XM_011516907.2 | XP_011515209.1 | 5, 6 | 1342 | ||
XM_011516909.1 | XP_011515211.1 | 5, 6 | 629 | ||
XM_011516910.1 | XP_011515212.1 | 6, 7 | 652 | ||
XM_017013174.1 | XP_016868663.1 | 5, 6 | 667 | ||
GenBank mRNA | AK092773.1 | - | 5, 6 | 592 | |
BC101659.1 | - | 5, 6 | 569 | 8069165 |