Gene Transcripts

Gene Symbol
ADCK5
Entrez Gene ID
203054
Gene Name
aarF domain containing kinase 5
Gene Aliases
Chromosome Location
Chr. 8: 144373762 - 144393242 on Build GRCh38
UniGene ID
Hs.283374
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_174922.4 NP_777582.4 10 1124
XM_006716527.2 XP_006716590.1 10 1114
XM_011516907.2 XP_011515209.1 10 1837
XM_011516909.1 XP_011515211.1 10 1124
XM_011516910.1 XP_011515212.1 11 1147
XM_017013174.1 XP_016868663.1 10 1162
GenBank mRNA AK092773.1 - 10 1087
BC064345.1 - 1 75
BC085013.1 - 5 434
BC101659.1 - 10 1064 8069165
BC143326.1 - 9 891
BU845476.1 - 5 432

View Full Product Details