Gene Transcripts

Gene Symbol
ADAM20
Entrez Gene ID
8748
Gene Name
ADAM metallopeptidase domain 20
Gene Aliases
Chromosome Location
Chr. 14: 70520817 - 70564742 on Build GRCh38
UniGene ID
Hs.177984
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003814.4 NP_003805.3 2 1787
XM_005268151.3 XP_005268208.1 2 10683
GenBank mRNA AF029899.1 - 1 1420
BC025378.1 - 2 1790 4827383

View Full Product Details