Gene Transcripts

Gene Symbol
ADAM28
Entrez Gene ID
10863
Gene Name
ADAM metallopeptidase domain 28
Gene Aliases
ADAM 28, MDC-L, MDCL, eMDC II, eMDCII
Chromosome Location
Chr. 8: 24294040 - 24359018 on Build GRCh38
UniGene ID
Hs.174030
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304351.1 NP_001291280.1 8 807
NM_014265.5 NP_055080.2 8 807
NM_021777.4 NP_068547.2 8 807
NR_130709.1 7 614
XM_005273380.3 XP_005273437.1 8 807
XM_005273382.1 XP_005273439.1 8 807
XM_006716273.3 XP_006716336.1 8 807
XM_006716274.1 XP_006716337.1 8 807
XM_011544367.2 XP_011542669.1 8 807
XM_011544368.2 XP_011542670.1 8 807
XM_011544369.2 XP_011542671.1 8 807
XM_011544370.2 XP_011542672.1 7 585
XM_017012974.1 XP_016868463.1 8 807
XM_017012975.1 XP_016868464.1 8 807
XM_017012976.1 XP_016868465.1 9 906
GenBank mRNA AF137334.1 - 8 736
AF137335.1 - 8 736
AJ242015.1 - 8 744
AK225921.1 - 777
AK293382.1 - 7 586
BC136478.1 - 8 723 9020467

View Full Product Details