Gene Transcripts

Gene Symbol
KLK15
Entrez Gene ID
55554
Gene Name
kallikrein related peptidase 15
Gene Aliases
ACO, HSRNASPH
Chromosome Location
Chr. 19: 50825289 - 50834337 on Build GRCh38
UniGene ID
Hs.567535
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277081.1 NP_001264010.1 5 918
NM_001277082.1 NP_001264011.1 4 781
NM_017509.3 NP_059979.2 5 921
NR_102274.1 5 800
XM_006723265.3 XP_006723328.1 6 1722
XM_011527085.2 XP_011525387.1 6 1718
XM_011527087.2 XP_011525389.1 5 1471
XM_011527089.2 XP_011525391.1 5 1592
GenBank mRNA AF303046.1 - 5 920
AY373373.1 - 4 1010
AY373374.1 - 4 873

View Full Product Details