Gene Transcripts
- Gene Symbol
- PTPN3
- Entrez Gene ID
- 5774
- Gene Name
- protein tyrosine phosphatase, non-receptor type 3
- Gene Aliases
- PTP-H1, PTPH1
- Chromosome Location
- Chr. 9: 109375694 - 109538395 on Build GRCh38
- UniGene ID
- Hs.436429
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001145368.1 |
NP_001138840.1
|
6, 7 |
533 |
|
NM_001145369.1 |
NP_001138841.1
|
1, 2 |
2582 |
|
NM_001145370.1 |
NP_001138842.1
|
1, 2 |
2582 |
|
NM_002829.3 |
NP_002820.3
|
6, 7 |
533 |
|
XM_006717197.3 |
XP_006717260.1
|
6, 7 |
642 |
|
XM_006717199.3 |
XP_006717262.1
|
6, 7 |
667 |
|
XM_006717201.3 |
XP_006717264.1
|
5, 6 |
474 |
|
XM_006717202.3 |
XP_006717265.1
|
3, 4 |
236 |
|
XM_006717203.3 |
XP_006717266.1
|
3, 4 |
195 |
|
XM_006717204.3 |
XP_006717267.1
|
6, 7 |
569 |
|
XM_011518888.2 |
XP_011517190.1
|
6, 7 |
603 |
|
XM_011518889.2 |
XP_011517191.1
|
6, 7 |
1273 |
|
XM_017014955.1 |
XP_016870444.1
|
6, 7 |
625 |
|
XM_017014956.1 |
XP_016870445.1
|
5, 6 |
481 |
|
XM_017014957.1 |
XP_016870446.1
|
6, 7 |
569 |
|
GenBank mRNA |
AK295875.1 |
- |
5, 6 |
453 |
|
AK304434.1 |
- |
6, 7 |
533 |
|
AK316066.1 |
- |
6, 7 |
533 |
|
BC126117.1 |
- |
5, 6 |
443 |
8991833 |
BC143848.1 |
- |
5, 6 |
443 |
9052363 |
BX648253.1 |
- |
1, 2 |
2583 |
|
BX648735.1 |
- |
1, 2 |
1017 |
|
DQ104439.1 |
- |
5, 6 |
421 |
|
M64572.1 |
- |
5, 6 |
444 |
|
View Full Product Details