Gene Transcripts
- Gene Symbol
- PTPRD
- Entrez Gene ID
- 5789
- Gene Name
- protein tyrosine phosphatase, receptor type D
- Gene Aliases
- HPTP, HPTPD, HPTPDELTA, PTPD, RPTPDELTA
- Chromosome Location
- Chr. 9: 8314246 - 10612840 on Build GRCh38
- UniGene ID
- Hs.446083
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB211400.1 |
- |
45, 46 |
6619 |
|
L38929.1 |
- |
36, 37 |
6061 |
|
View Full Product Details