Gene Transcripts

Gene Symbol
TRPV2
Entrez Gene ID
51393
Gene Name
transient receptor potential cation channel subfamily V member 2
Gene Aliases
VRL, VRL-1, VRL1
Chromosome Location
Chr. 17: 16415542 - 16437003 on Build GRCh38
UniGene ID
Hs.279746
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016113.4 NP_057197.2 5, 6 1319
XM_005256676.1 XP_005256733.1 5, 6 1319
XM_005256677.1 XP_005256734.1 5, 6 1319
XM_005256678.4 XP_005256735.1 5, 6 1319
XM_006721541.3 XP_006721604.1 5, 6 1319
XM_006721543.3 XP_006721606.1 5, 6 1319
XM_011523922.1 XP_011522224.1 5, 6 1319
XM_011523923.1 XP_011522225.1 5, 6 1319
XM_017024730.1 XP_016880219.1 5, 6 1319
XM_017024731.1 XP_016880220.1 5, 6 1319
XM_017024732.1 XP_016880221.1 5, 6 1319
GenBank mRNA AF103906.1 - 5, 6 1085
AF129112.1 - 4, 5 955
AJ487963.1 - 4, 5 924
AK289705.1 - 5, 6 1319
BC018926.2 - 5, 6 1287 4298484
BC051305.1 - 5, 6 1275 4870010

View Full Product Details