Gene Transcripts

Gene Symbol
INPP4B
Entrez Gene ID
8821
Gene Name
inositol polyphosphate-4-phosphatase type II B
Gene Aliases
Chromosome Location
Chr. 4: 142023160 - 142846535 on Build GRCh38
UniGene ID
Hs.531403
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101669.1 NP_001095139.1 7 858
NM_003866.2 NP_003857.2 8 957
XM_011532391.2 XP_011530693.1 7 942
XM_011532392.2 XP_011530694.1 7 708
XM_017008794.1 XP_016864283.1 7 942
XM_017008795.1 XP_016864284.1 7 942
XM_017008796.1 XP_016864285.1 7 942
GenBank mRNA AY753912.1 - 4 362
BC005274.1 - 7 633
BC072447.1 - 6 765
BC110918.1 - 6 715 6160633
BC144255.1 - 4 413
BX649090.1 - 8 798
U96922.1 - 4 483

View Full Product Details