Gene Transcripts

Gene Symbol
PKD1L2
Entrez Gene ID
114780
Gene Name
polycystin 1 like 2 (gene/pseudogene)
Gene Aliases
PC1L2
Chromosome Location
Chr. 16: 81100879 - 81220370 on Build GRCh38
UniGene ID
Hs.413525
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001076780.1 NP_001070248.1 1 186
NM_052892.3 NP_443124.3 1 186
NR_126532.1 1 186
GenBank mRNA AY164483.1 - 1 186

View Full Product Details