Gene Transcripts

Gene Symbol
KCNK18
Entrez Gene ID
338567
Gene Name
potassium two pore domain channel subfamily K member 18
Gene Aliases
K2p18.1, MGR13, TRESK, TRESK-2, TRESK2, TRIK
Chromosome Location
Chr. 10: 117197489 - 117210299 on Build GRCh38
UniGene ID
Hs.449650
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181840.1 NP_862823.1 3 1090
GenBank mRNA AB087138.1 - 3 1090

View Full Product Details