Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001317237.1 | NP_001304166.1 | 10 | 1817 | |
NM_001908.4 | NP_001899.1 | 10 | 1848 | ||
NM_147780.3 | NP_680090.1 | 12 | 2010 | ||
NM_147781.3 | NP_680091.1 | 11 | 1967 | ||
NM_147782.3 | NP_680092.1 | 11 | 1936 | ||
NM_147783.3 | NP_680093.1 | 11 | 1922 | ||
XM_006716244.2 | XP_006716307.1 | 12 | 2034 | ||
XM_006716245.2 | XP_006716308.1 | 10 | 2138 | ||
XM_011543812.2 | XP_011542114.1 | 12 | 1943 | ||
XM_017013097.1 | XP_016868586.1 | 11 | 1924 | ||
XM_017013098.1 | XP_016868587.1 | 11 | 1924 | ||
XM_017013099.1 | XP_016868588.1 | 12 | 1966 | ||
XM_017013100.1 | XP_016868589.1 | 11 | 1868 | ||
XM_017013101.1 | XP_016868590.1 | 7 | 1391 | ||
GenBank mRNA | AK075393.1 | - | 10 | 1824 | |
AK092070.1 | - | 11 | 1859 | ||
AK097384.1 | - | 5 | 1778 | ||
AK128472.1 | - | 1 | 2441 | ||
AK130184.1 | - | 10 | 1785 | ||
BC010240.1 | - | 11 | 1865 | 4154307 | |
BC095408.1 | - | 10 | 1785 | 30334082 | |
BX647765.1 | - | 10 | 1708 | ||
BX647952.1 | - | 10 | 1788 | ||
BX648239.1 | - | 5 | 7559 | ||
L16510.1 | - | 11 | 1843 | ||
M14221.1 | - | 11 | 1850 |