Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318006.1 | NP_001304935.1 | 4 | 1039 | |
NM_001318007.1 | NP_001304936.1 | 4 | 933 | ||
NM_001318008.1 | NP_001304937.1 | 3 | 837 | ||
NM_024792.2 | NP_079068.1 | 5 | 1135 | ||
GenBank mRNA | AF477201.1 | - | 5 | 1123 | |
AK025935.1 | - | 5 | 1074 | ||
AK125146.1 | - | 4 | 1340 | ||
BC009729.1 | - | 2 | 628 | ||
BC026023.1 | - | 4 | 1001 | 4548729 | |
BX389422.1 | - | 3 | 680 |