Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031474.2 | NP_113662.1 | 6 | 951 | |
XM_011521031.2 | XP_011519333.1 | 7 | 893 | ||
XM_017020000.1 | XP_016875489.1 | 4 | 1524 | ||
GenBank mRNA | AK054740.1 | - | 2 | 162 | |
AK098063.1 | - | 6 | 922 | ||
AL136557.1 | - | 6 | 919 | ||
BC036063.1 | - | 6 | 920 | 5276999 | |
BC131562.1 | - | 6 | 908 | 40109200 |