Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001301175.1 | NP_001288104.1 | 13 | 1405 | |
NM_032663.4 | NP_116052.2 | 10 | 1031 | ||
XM_005253962.3 | XP_005254019.1 | 10 | 1014 | ||
XM_005253965.4 | XP_005254022.1 | 9 | 842 | ||
XM_006719653.3 | XP_006719716.1 | 11 | 1104 | ||
XM_011538894.2 | XP_011537196.1 | 8 | 735 | ||
XM_017020048.1 | XP_016875537.1 | 10 | 1026 | ||
XM_017020049.1 | XP_016875538.1 | 10 | 1014 | ||
XM_017020050.1 | XP_016875539.1 | 10 | 1126 | ||
XM_017020051.1 | XP_016875540.1 | 11 | 982 | ||
XM_017020052.1 | XP_016875541.1 | 10 | 1028 | ||
XM_017020053.1 | XP_016875542.1 | 9 | 758 | ||
XM_017020054.1 | XP_016875543.1 | 8 | 633 | ||
GenBank mRNA | AJ586136.1 | - | 10 | 922 | |
AK027820.1 | - | 10 | 996 | ||
AK097830.1 | - | 13 | 1405 | ||
AL834278.1 | - | 1 | 194 | ||
BC004868.2 | - | 10 | 1021 | 3833242 | |
BC022094.2 | - | 8 | 573 | ||
CN367077.1 | - | 4 | 295 |