Gene Transcripts

Gene Symbol
CNN3
Entrez Gene ID
1266
Gene Name
calponin 3
Gene Aliases
Chromosome Location
Chr. 1: 94896949 - 94927223 on Build GRCh38
UniGene ID
Hs.483454
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286055.1 NP_001272984.1 2 418
NM_001286056.1 NP_001272985.1 2 208
NM_001839.4 NP_001830.1 2 418
XM_017000245.1 XP_016855734.1 4 2830
GenBank mRNA AK294141.1 - 2 169
AK298145.1 - 2 338
AK312370.1 - 2 305
AK316398.1 - 2 184
BC025372.2 - 2 302 3505668
BU149686.1 - 2 354
CR541848.1 - 2 89
DC422152.1 - 2 374
DR157009.1 - 2 341
S80562.1 - 2 172

View Full Product Details