Gene Transcripts

Gene Symbol
RCE1
Entrez Gene ID
9986
Gene Name
Ras converting CAAX endopeptidase 1
Gene Aliases
FACE2, RCE1A, RCE1B
Chromosome Location
Chr. 11: 66843412 - 66846532 on Build GRCh38
UniGene ID
Hs.654972
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001032279.1 NP_001027450.1 8 1452
NM_005133.2 NP_005124.1 8 1456
GenBank mRNA AF121951.1 - 8 1457
BC052622.1 - 8 1413

View Full Product Details