Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001862.2 | NP_001853.2 | 3 | 258 | |
GenBank mRNA | BC006229.2 | - | 3 | 258 | |
BT006742.1 | - | 3 | 229 | ||
CR541727.1 | - | 3 | 229 | ||
CR541753.1 | - | 3 | 229 | ||
M19961.1 | - | 3 | 250 |