Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001172632.1 | NP_001166103.1 | 5 | 1249 | |
NM_001172633.1 | NP_001166104.1 | 5 | 1273 | ||
NM_002543.3 | NP_002534.1 | 6 | 1389 | ||
GenBank mRNA | AB010710.1 | - | 6 | 1336 | |
AF035776.1 | - | 6 | 1321 | ||
AX429248.1 | - | 6 | 1336 | ||
BC022295.1 | - | 6 | 1330 | 4722086 |