Gene Transcripts

Gene Symbol
RBP3
Entrez Gene ID
5949
Gene Name
retinol binding protein 3
Gene Aliases
D10S64, D10S65, D10S66, IRBP, RBPI, RP66
Chromosome Location
Chr. 10: 47348371 - 47357875 on Build GRCh38
UniGene ID
Hs.591928
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002900.2 NP_002891.1 1 931
GenBank mRNA AB593121.1 - 1 923
BC039844.1 - 1 921 5578501
J03912.1 - 1 814
M22453.1 - 1 932

View Full Product Details