Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039573.2 | NP_001034662.2 | 11 | 1455 | |
NM_001143998.1 | NP_001137470.1 | 11 | 1455 | ||
NM_001143999.1 | NP_001137471.1 | 11 | 1451 | ||
NM_001144001.1 | NP_001137473.1 | 9 | 1243 | ||
NM_001204408.1 | NP_001191337.1 | 13 | 1800 | ||
NM_001204410.1 | NP_001191339.1 | 12 | 1493 | ||
NM_003003.3 | NP_002994.3 | 11 | 1402 | ||
GenBank mRNA | AB208811.1 | - | 11 | 1434 | |
AK290913.1 | - | 11 | 1348 | ||
AK293206.1 | - | 9 | 1243 | ||
BC136523.1 | - | 11 | 1290 | 9020513 | |
BC136525.1 | - | 11 | 1290 | 9020515 | |
BC142979.1 | - | 11 | 1346 | 8860414 | |
BX537448.1 | - | 11 | 1356 | ||
D67029.1 | - | 11 | 1438 | ||
FM995495.1 | - | 13 | 1694 | ||
FM995496.1 | - | 12 | 1491 | ||
FM995497.1 | - | 12 | 1590 |