Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003742.2 | NP_003733.2 | 21 | 2623 | |
XM_006712817.3 | XP_006712880.1 | 21 | 2735 | ||
XM_011512077.2 | XP_011510379.1 | 21 | 2795 | ||
XM_011512078.2 | XP_011510380.1 | 21 | 2795 | ||
XM_011512080.2 | XP_011510382.1 | 21 | 2798 | ||
XM_011512081.2 | XP_011510383.1 | 8 | 1186 | ||
XM_017005165.1 | XP_016860654.1 | 21 | 2797 | ||
XM_017005166.1 | XP_016860655.1 | 15 | 1921 | ||
XM_017005167.1 | XP_016860656.1 | 11 | 1479 | ||
GenBank mRNA | AF091582.1 | - | 21 | 2623 | |
AF136523.1 | - | 21 | 2624 | ||
AK302540.1 | - | 8 | 1074 | ||
AK303050.1 | - | 8 | 1186 |