Gene Transcripts

Gene Symbol
MECOM
Entrez Gene ID
2122
Gene Name
MDS1 and EVI1 complex locus
Gene Aliases
AML1-EVI-1, EVI1, MDS1, MDS1-EVI1, PRDM3, RUSAT2
Chromosome Location
Chr. 3: 169083499 - 169663781 on Build GRCh38
UniGene ID
Hs.744090
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC069498.1 - 3 725
CR541866.1 - 3 473
CR541886.1 - 3 473
U43292.1 - 3 780

View Full Product Details