Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001916.4 | NP_001907.2 | 4 | 578 | |
XM_017013102.1 | XP_016868591.1 | 3 | 1111 | ||
GenBank mRNA | AK026633.1 | - | 4 | 536 | |
AK225796.1 | - | 3 | 1179 | ||
BC001006.1 | - | 4 | 524 | 2821836 | |
BC015616.2 | - | 4 | 518 | 4840922 | |
BC020566.1 | - | 4 | 517 | 4157475 | |
BC024650.1 | - | 4 | 485 | ||
BG328536.1 | - | 4 | 578 | ||
BT019798.1 | - | 4 | 513 | ||
CN369438.1 | - | 4 | 520 | ||
CR541674.1 | - | 4 | 513 | ||
M16597.1 | - | 3 | 219 | ||
X06994.1 | - | 4 | 521 |