Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001927.3 | NP_001918.3 | 2 | 697 | |
GenBank mRNA | AF055081.1 | - | 2 | 611 | |
AF055082.1 | - | 2 | 611 | ||
AF055083.1 | - | 2 | 611 | ||
AF137053.1 | - | 2 | 611 | ||
AF167579.1 | - | 2 | 611 | ||
AF486807.1 | - | 2 | 611 | ||
AF487828.1 | - | 2 | 611 | ||
AF521879.1 | - | 2 | 611 | ||
AF527578.1 | - | 2 | 611 | ||
AK097038.1 | - | 3 | 637 | ||
AK098332.1 | - | 3 | 491 | ||
AK123787.1 | - | 3 | 436 | ||
AK300654.1 | - | 692 | |||
AY083345.1 | - | 2 | 611 | ||
AY125465.1 | - | 2 | 611 | ||
BC032116.2 | - | 2 | 709 | 4905678 | |
DQ104335.1 | - | 2 | 611 | ||
DQ104336.1 | - | 2 | 611 | ||
DQ104337.1 | - | 2 | 611 | ||
EF617312.1 | - | 2 | 691 | ||
JX114780.1 | - | 2 | 611 | ||
U59167.1 | - | 2 | 688 |