Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001003688.1 | NP_001003688.1 | 5 | 1031 | |
NM_005900.2 | NP_005891.1 | 5 | 1207 | ||
XM_005262992.3 | XP_005263049.1 | 5 | 1000 | ||
XM_006714217.2 | XP_006714280.1 | 5 | 1062 | ||
XM_011531961.1 | XP_011530263.1 | 5 | 1022 | ||
XM_011531962.1 | XP_011530264.1 | 5 | 1067 | ||
XM_011531963.1 | XP_011530265.1 | 5 | 1189 | ||
XM_011531964.1 | XP_011530266.1 | 5 | 1219 | ||
GenBank mRNA | AK096624.1 | - | 1 | 2542 | |
AK293055.1 | - | 5 | 1333 | ||
BC001878.1 | - | 5 | 1132 | 3546771 | |
BT007386.1 | - | 4 | 791 | ||
U54826.1 | - | 5 | 1031 | ||
U57456.1 | - | 4 | 943 | ||
U59423.1 | - | 5 | 1223 | ||
U59912.1 | - | 5 | 1032 |