Gene Transcripts
- Gene Symbol
- NFRKB
- Entrez Gene ID
- 4798
- Gene Name
- nuclear factor related to kappaB binding protein
- Gene Aliases
- INO80G
- Chromosome Location
- Chr. 11: 129863773 - 129895596 on Build GRCh38
- UniGene ID
- Hs.530539
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001143835.1 |
NP_001137307.1
|
18, 19 |
2063 |
|
NM_006165.3 |
NP_006156.2
|
16, 17 |
2058 |
|
XM_005271567.3 |
XP_005271624.1
|
18, 19 |
2099 |
|
XM_005271568.3 |
XP_005271625.1
|
17, 18 |
2014 |
|
XM_011542850.2 |
XP_011541152.1
|
19, 20 |
2136 |
|
XM_011542851.2 |
XP_011541153.1
|
18, 19 |
2118 |
|
XM_011542852.2 |
XP_011541154.1
|
17, 18 |
1956 |
|
XM_017017793.1 |
XP_016873282.1
|
17, 18 |
2010 |
|
XM_017017794.1 |
XP_016873283.1
|
17, 18 |
1974 |
|
XM_017017795.1 |
XP_016873284.1
|
19, 20 |
2100 |
|
XM_017017796.1 |
XP_016873285.1
|
17, 18 |
1978 |
|
XM_017017797.1 |
XP_016873286.1
|
17, 18 |
2010 |
|
XM_017017798.1 |
XP_016873287.1
|
18, 19 |
2063 |
|
XM_017017799.1 |
XP_016873288.1
|
19, 20 |
2210 |
|
XM_017017800.1 |
XP_016873289.1
|
19, 20 |
2150 |
|
XM_017017801.1 |
XP_016873290.1
|
4, 5 |
1110 |
|
GenBank mRNA |
BC063280.1 |
- |
18, 19 |
2009 |
|
X80878.1 |
- |
16, 17 |
2067 |
|
View Full Product Details