Gene Transcripts
- Gene Symbol
- SSR4
- Entrez Gene ID
- 6748
- Gene Name
- signal sequence receptor subunit 4
- Gene Aliases
- CDG1Y, TRAPD
- Chromosome Location
- Chr. X: 153794175 - 153798512 on Build GRCh38
- UniGene ID
- Hs.409223
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001204526.1 |
NP_001191455.1
|
5, 6 |
510 |
|
NM_001204527.1 |
NP_001191456.1
|
5, 6 |
533 |
|
NM_006280.2 |
NP_006271.1
|
4, 5 |
483 |
|
NR_037927.1 |
|
6, 7 |
704 |
|
XM_011531186.1 |
XP_011529488.1
|
5, 6 |
392 |
|
XM_011531187.1 |
XP_011529489.1
|
5, 6 |
392 |
|
XM_017029756.1 |
XP_016885245.1
|
5, 6 |
1281 |
|
XM_017029757.1 |
XP_016885246.1
|
5, 6 |
1016 |
|
GenBank mRNA |
AK057117.1 |
- |
3, 4 |
2066 |
|
AK094395.1 |
- |
4, 5 |
2186 |
|
AK290493.1 |
- |
4, 5 |
389 |
|
BC003371.1 |
- |
4, 5 |
363 |
2900758 |
BE791319.1 |
- |
4, 5 |
405 |
|
BG165852.1 |
- |
5, 6 |
517 |
|
BP202705.1 |
- |
5, 6 |
472 |
|
BT007192.1 |
- |
4, 5 |
359 |
|
CD672615.1 |
- |
5, 6 |
575 |
|
DB238237.1 |
- |
4, 5 |
482 |
|
X90583.1 |
- |
4, 5 |
389 |
|
Z69043.1 |
- |
4, 5 |
408 |
|
View Full Product Details