Gene Transcripts
- Gene Symbol
- EYA3
- Entrez Gene ID
- 2140
- Gene Name
- EYA transcriptional coactivator and phosphatase 3
- Gene Aliases
- Chromosome Location
- Chr. 1: 27970344 - 28088671 on Build GRCh38
- UniGene ID
- Hs.185774
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001282560.1 |
NP_001269489.1
|
4, 5 |
343 |
|
NM_001282561.1 |
NP_001269490.1
|
4, 5 |
343 |
|
NM_001990.3 |
NP_001981.2
|
4, 5 |
343 |
|
NR_104214.1 |
|
4, 5 |
343 |
|
XM_006710449.3 |
XP_006710512.1
|
5, 6 |
379 |
|
XM_011540999.2 |
XP_011539301.1
|
5, 6 |
382 |
|
XM_011541001.2 |
XP_011539303.1
|
4, 5 |
340 |
|
XM_011541003.2 |
XP_011539305.1
|
5, 6 |
380 |
|
XM_011541004.2 |
XP_011539306.1
|
5, 6 |
377 |
|
XM_017000652.1 |
XP_016856141.1
|
5, 6 |
383 |
|
XM_017000653.1 |
XP_016856142.1
|
6, 7 |
503 |
|
XM_017000654.1 |
XP_016856143.1
|
4, 5 |
340 |
|
XM_017000655.1 |
XP_016856144.1
|
5, 6 |
496 |
|
XM_017000656.1 |
XP_016856145.1
|
5, 6 |
381 |
|
XM_017000657.1 |
XP_016856146.1
|
4, 5 |
373 |
|
GenBank mRNA |
AK289805.1 |
- |
4, 5 |
302 |
|
AK295745.1 |
- |
4, 5 |
337 |
|
AL603098.1 |
- |
4, 5 |
326 |
|
BC029500.1 |
- |
4, 5 |
303 |
5272319 |
BC041667.1 |
- |
4, 5 |
270 |
5743906 |
BX648945.1 |
- |
4, 5 |
312 |
|
DC416048.1 |
- |
4, 5 |
343 |
|
View Full Product Details