Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001007267.2 | NP_001007268.1 | 4 | 1013 | |
NM_001195641.1 | NP_001182570.1 | 4 | 1013 | ||
NM_007366.4 | NP_031392.3 | 4 | 1013 | ||
XM_005246392.3 | XP_005246449.1 | 4 | 901 | ||
XM_011510820.2 | XP_011509122.1 | 4 | 901 | ||
XM_017003598.1 | XP_016859087.1 | 4 | 901 | ||
XM_017003599.1 | XP_016859088.1 | 4 | 901 | ||
XM_017003601.1 | XP_016859090.1 | 4 | 1300 | ||
GenBank mRNA | BC140823.1 | - | 4 | 823 | |
BC144631.1 | - | 4 | 823 | 9053162 | |
U17033.1 | - | 4 | 1013 | ||
U17034.1 | - | 4 | 1013 |