Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001206615.1 | NP_001193544.1 | 8 | 1599 | |
NM_001206616.1 | NP_001193545.1 | 9 | 1669 | ||
NM_012153.5 | NP_036285.2 | 9 | 1668 | ||
XM_005252857.1 | XP_005252914.1 | 9 | 1666 | ||
XM_005252860.1 | XP_005252917.1 | 9 | 1522 | ||
XM_005252861.1 | XP_005252918.1 | 8 | 1600 | ||
XM_005252862.1 | XP_005252919.1 | 8 | 1469 | ||
XM_011519983.1 | XP_011518285.1 | 9 | 1694 | ||
XM_011519984.1 | XP_011518286.1 | 10 | 1774 | ||
XM_011519985.1 | XP_011518287.1 | 9 | 1559 | ||
XM_017017534.1 | XP_016873023.1 | 8 | 1597 | ||
GenBank mRNA | AF124438.1 | - | 8 | 1500 | |
AF124439.1 | - | 9 | 1569 | ||
AF170583.1 | - | 9 | 1578 | ||
AF203977.1 | - | 9 | 1589 | ||
AF212848.1 | - | 9 | 1608 |