Gene Transcripts
- Gene Symbol
- COMMD5
- Entrez Gene ID
- 28991
- Gene Name
- COMM domain containing 5
- Gene Aliases
- HCARG, HT002
- Chromosome Location
- Chr. 8: 144837978 - 144853578 on Build GRCh38
- UniGene ID
- Hs.493218
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK023070.1 |
- |
3 |
1514 |
|
View Full Product Details