Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001304495.1 | NP_001291424.1 | 4 | 1835 | |
NM_171982.4 | NP_741983.2 | 6 | 1954 | ||
XM_005273452.3 | XP_005273509.1 | 5 | 1926 | ||
XM_011544453.1 | XP_011542755.1 | 7 | 1982 | ||
GenBank mRNA | AB029021.1 | - | 6 | 1904 | |
AF492463.1 | - | 6 | 1952 | ||
BC018337.1 | - | 6 | 2064 | ||
BC069226.1 | - | 6 | 1896 | 6428667 |