Gene Transcripts

Gene Symbol
AASDHPPT
Entrez Gene ID
60496
Gene Name
aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase
Gene Aliases
AASD-PPT, ACPS, CGI-80, LYS2, LYS5
Chromosome Location
Chr. 11: 106077565 - 106098692 on Build GRCh38
UniGene ID
Hs.524009
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015423.2 NP_056238.2 6 2779
GenBank mRNA AB072896.1 - 1 1717
AF302110.1 - 6 2798
AL050073.1 - 5 2339
BX537665.1 - 2 5054

View Full Product Details